4-Toluoyl chloride 98%
Leverantör: Thermo Scientific
Fara
Synonyms: p-Toluoyl chloride, 4-Methylbenzene-1-carbonyl chloride, p-Methylbenzoyl chloride, 4-Methylbenzoyl chloride
147980050 147982500 147980010
ACRO147980050EA 550 SEK
ACRO147980050 ACRO147982500 ACRO147980010
4-Toluoyl chloride 98%
4-Toluoyl chloride
Formel: C₈H₇ClO
MW: 154,6 g/mol
Kokpunkt: 225…227 °C (760 mmHg)
Smältpunkt: –2 °C
Densitet: 1,16 g/cm³
Flampunkt: 82 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000696
CAS nummer: 874-60-2
EINECS: 212-864-8
UN: 3265
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5525 to 1.5545 (20°C, 589 nm)
Titration RCOCl ≥97.5 %