4-Ethylbenzoyl chloride 97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 4-Ethylbenzene-1-carbonyl chloride, 4-Ethylbenzoylchloride
305730050 305730250
ACRO305730050EA 845 SEK
ACRO305730050 ACRO305730250
4-Ethylbenzoyl chloride 97%
4-Ethylbenzoyl chloride
Formel: C₉H₉ClO
Kokpunkt: 235…236 °C (760 mmHg)
Smältpunkt: 42…43 °C
Densitet: 1,14 g/cm³
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000697
CAS nummer: 16331-45-6
EINECS: 240-404-6
UN: 3265
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5465 to 1.5485 (20°C, 589 nm)
GC ≥96.0 %