2'-Hydroxyacetophenone 99%
Leverantör: Thermo Scientific
Varning
Synonyms: o-Hydroxyacetophenone, 2-Acetophenol, 2-Hydroxyphenyl Methyl Ketone, 1-(2-Hydroxyphenyl)ethan-1-one, o-Acetylphenol, 1-(2-Hydroxyphenyl)-1-oxoethane, 1-Ethanoyl-2-hydroxybenzol, 2-Acetylphenol
120931000 120935000
ACRO120931000EA 555 SEK
ACRO120931000 ACRO120935000
2'-Hydroxyacetophenone 99%
2'-Hydroxyacetophenone
Formel: C₈H₈O₂
MW: 136,15 g/mol
Kokpunkt: 92…93 °C (12 mmHg)
Smältpunkt: 4…6 °C
Densitet: 1,131 g/cm³
Flampunkt: 112 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002219
CAS nummer: 118-93-4
EINECS: 204-288-0
Order Now
Specifikation testresultat
Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5580 to 1.5600 (20°C, 589 nm)
GC ≥98.5 %