3',4'-Dimethoxyacetophenone ≥98%
Leverantör: Thermo Scientific
Synonyms: 1-(3,4-Dimethoxyphenyl)ethan-1-one, 3,4-Dimethoxyacetophenone, 1-(3,4-Dimethoxyphenyl)ethanone
L02737.18 L02737.30
L02737.18EA 805 SEK
L02737.18 L02737.30
3',4'-Dimethoxyacetophenone ≥98%
3',4'-Dimethoxyacetophenone
Purity: 98+% CAS No.: 1131-62-0 Molecular Formula: C10H12O3 Formula Weight: 180.20 Flash Point: 110°(230°F) Odour: Odourless Boiling Point: 286-288° Beilstein Reference: 781213 MDL No.: MFCD00008737
Formel: C₁₀H₁₂O₃
MW: 180,2 g/mol
Kokpunkt: 286…288 °C
Smältpunkt: 47…51 °C
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008737
CAS nummer: 1131-62-0
EINECS: 214-468-0