ω,ω-Diethoxyacetophenone 96%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 2,2-Diethoxyacetophenone, 2,2-Diethoxy-1-phenylethan-1-one
210620050 210621000
ACRO210620050EA 380 SEK
ACRO210620050 ACRO210621000
ω,ω-Diethoxyacetophenone 96%
ω,ω-Diethoxyacetophenone
Formel: C₁₂H₁₆O₃
Kokpunkt: 131…134 °C (10 mmHg)
Densitet: 1,03 g/cm³
Flampunkt: 128 °C
MDL Number: MFCD00009659
CAS nummer: 6175-45-7
EINECS: 228-220-4
Order Now
Specifikation testresultat
Appearance Clear colorless to dark yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4980 to 1.5010 (20°C, 589 nm)
GC ≥95 %