4'-Methoxypropiophenone ≥99%
Leverantör: Thermo Fisher Scientific
Varning
A14619.14 A14619.22 A14619.36
A14619.14EA 1200 SEK
A14619.14 A14619.22 A14619.36
4'-Methoxypropiophenone ≥99%
4'-Methoxypropiophenone
Purity: 99% CAS No.: 121-97-1 Molecular Formula: C10H12O2 Formula Weight: 164.20 Flash Point: 62°(143°F) Refractive Index: 1.5465 Melting Point: 27-29° Boiling Point: 273-275° Beilstein Reference: 907733 MDL No.: MFCD00009310
Formel: C₁₀H₁₂O₂
MW: 164.2 g/mol
Kokpunkt: 273…275 °C
Smältpunkt: 27…29 °C
Densitet: 1.070
Flampunkt: 62 °C (143 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009310
CAS nummer: 121-97-1
EINECS: 204-512-7