2'-Aminoacetophenone 97%
Leverantör: Thermo Fisher Scientific
Varning
400620100 400620050 400620500
ACRO400620100EA 1225 SEK
ACRO400620100 ACRO400620050 ACRO400620500
2'-Aminoacetophenone 97%
2'-Aminoacetophenone
Formel: C₈H₉NO
Kokpunkt: 70…71 °C (3 mmHg)
Smältpunkt: 20 °C
Densitet: 1,12 g/cm³ (20 °C)
Flampunkt: 112 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00007717
CAS nummer: 551-93-9
EINECS: 209-002-8
Merck Index: 13,00412
Order Now
Specifikation testresultat
Appearance Yellow to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.6130 to 1.6170 (20°C, 589 nm)
GC ≥96.0 %