3-Acetylbenzoic acid ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 3'-Acetophenonecarboxylic acid, 3-Acetylbenzoic Acid
B25616.03 B25616.06
B25616.03EA 2225 SEK
B25616.03 B25616.06
3-Acetylbenzoic acid ≥98%
3-Acetylbenzoic acid
Purity: 98% CAS No.: 586-42-5 Molecular Formula: C9H8O3 Formula Weight: 164.16 Odour: Odourless Melting Point: 162-166° MDL No.: MFCD00045847
Formel: C₉H₈O₃
Kokpunkt: 343 °C (1013 hPa)
Smältpunkt: 162…166 °C
Densitet: 1,23 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00045847
CAS nummer: 586-42-5