6-Klor-m-toluidin 99%, Sigma-Aldrich®
Leverantör: SIGMA ALDRICH MICROSCOPY
Varning
225118-25G 225118-5G
225118-25GEA 5200 SEK
225118-25G 225118-5G
6-Klor-m-toluidin 99%, Sigma-Aldrich®
6-Klor-m-toluidin
Formel: C₇H₈ClN
MW: 141.6 g/mol
Kokpunkt: 228…230 °C (1013 hPa)
Smältpunkt: 30
Flampunkt: 107 °C (closed cup)
Storage Temperature: Room Temperature
MDL Number: MFCD00007674
CAS nummer: 95-81-8
EINECS: 202-454-7
UN: 2239
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc4f3e7d19195ef2f0c03e1a9bd73e315a1480ae&vid=9fbb7000f8a3094cd4d37543991d289a70579707&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to tan solid, crystals, powder, crystalline powder and/or chunks
Infrared spectrum Conforms to structure
98.5% (minimum) 98.5 - 101.5% (with AgNO₃ after O₂ combustion)