2-Klor-p-toluidin 98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 2-Chloro-4-methylaniline, 4-Amino-3-chlorotoluene
154670250 154670050
ACRO154670250EA 1125 SEK
ACRO154670250 ACRO154670050
2-Klor-p-toluidin 98%
2-Klor-p-toluidin
Formel: C₇H₈ClN
MW: 141.6 g/mol
Kokpunkt: 223…225 °C (760 mmHg)
Smältpunkt: 7 °C
Densitet: 1.151 g/cm³
Flampunkt: 99 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007666
CAS nummer: 615-65-6
EINECS: 210-440-7
UN: 3429
ADR: 6.1 III
Order Now
Specifikation testresultat
Appearance Clear yellow to light brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5740 to 1.5760 (20°C, 589 nm)
GC ≥97.5 %
NMR (1H-NMR) Consistent with structure