3-Chlorobenzylamine 98%
Leverantör: Thermo Scientific
Fara
Synonyms: (3-Chlorophenyl)methanamine
297570050 297570250
ACRO297570050EA 995 SEK
ACRO297570050 ACRO297570250
3-Chlorobenzylamine 98%
3-Chlorobenzylamine
Formel: C₇H₈ClN
MW: 141,6 g/mol
Kokpunkt: 110…112 °C (17 mmHg)
Densitet: 1,15 g/cm³
Flampunkt: 98 °C
MDL Number: MFCD00040752
CAS nummer: 4152-90-3
EINECS: 223-985-0
UN: 2735
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5575 to 1.5605 (20°C, 589 nm)
GC ≥97.5 %