2-Chloro-4-(trifluoromethyl)aniline ≥98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 4-Amino-3-chloro-α,α,α-trifluorotoluene, 2-Chloro-α,α,α-trifluoro-p-toluidine, 4-Amino-3-chlorobenzotrifluoride
A12242.06 A12242.14 A12242.22
A12242.06EA 1225 SEK
A12242.06 A12242.14 A12242.22
2-Chloro-4-(trifluoromethyl)aniline ≥98%
2-Chloro-4-(trifluoromethyl)aniline
Purity: 98% CAS No.: 39885-50-2 Molecular Formula: C7H5ClF3N Formula Weight: 195.57 Flash Point: 103°(217°F) Refractive Index: 1.5015 Boiling Point: 214-218° Beilstein Reference: 4179703 UN No.: 2810 MDL No.: MFCD00042563
Formel: C₇H₅ClF₃N
Kokpunkt: 214…218 °C
Densitet: 1,434
Flampunkt: 103 °C (217 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00042563
CAS nummer: 39885-50-2
EINECS: 254-674-8
UN: 2810
ADR: 6.1,III