3'-Chloroacetophenone 98%
Leverantör: Thermo Fisher Scientific
Varning
134840250 134841000
ACRO134840250EA 1600 SEK
ACRO134840250 ACRO134841000
3'-Chloroacetophenone 98%
3'-Chloroacetophenone
Formel: C₈H₇ClO
Kokpunkt: 241 °C (1013 hPa)
Densitet: 1,19 g/cm³
Flampunkt: 105 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000593
CAS nummer: 99-02-5
EINECS: 202-721-8
ADR: 6.1,II
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5480 to 1.5500 (20°C, 589 nm)
GC ≥97.5 %