β-Chloropropiophenone 96%
Leverantör: Thermo Scientific
Synonyms: 3-Chloropropiophenone
A12290.09 A12290.18 A12290.22
A12290.09EA 1275 SEK
A12290.09 A12290.18 A12290.22
β-Chloropropiophenone 96%
β-Chloropropiophenone
Purity: 96% CAS No.: 936-59-4 Molecular Formula: C9H9ClO Formula Weight: 168.62 Flash Point: 110°(230°F) Melting Point: 46-50° Boiling Point: 113-115°/4mm Beilstein Reference: 2043580 MDL No.: MFCD00000990
Formel: (C₆H₅)C(O)CH₂CH₂Cl
MW: 168,62 g/mol
Kokpunkt: 113…115 °C (4 mmHg)
Smältpunkt: 46…50 °C
Densitet: 1,181 g/cm³ (20 °C)
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000990
CAS nummer: 936-59-4
EINECS: 213-317-6