4-Bromobenzylamine 97+%
Leverantör: Thermo Scientific
Fara
350530050 350530250
ACRO350530050EA 5575 SEK
ACRO350530050 ACRO350530250
4-Bromobenzylamine 97+%
4-Bromobenzylamine
Formel: C₇H₈BrN
MW: 186,05 g/mol
Smältpunkt: 25 °C
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00047931
CAS nummer: 3959-07-7
EINECS: 223-559-4
UN: 3259
ADR: 8,III
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid or solid
Infrared spectrum Conforms
Refractive index 1.5860 to 1.5890 (20°C, 589 nm)
GC ≥97.0 %