5-Bromo-α,α,α-trifluoro-2-nitrotoluene ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Bromo-4-nitro-3-(trifluoromethyl)benzene, 5-Bromo-2-nitrobenzotrifluoride
A17634.06 A17634.14 A17634.22
A17634.06EA 780 SEK
A17634.06 A17634.14 A17634.22
5-Bromo-α,α,α-trifluoro-2-nitrotoluene ≥98%
5-Bromo-α,α,α-trifluoro-2-nitrotoluene
Purity: 98+% CAS No.: 344-38-7 Molecular Formula: C7H3BrF3NO2 Formula Weight: 270.01 Flash Point: 110°(230°F) Refractive Index: 1.5180 Melting Point: 32-35° Boiling Point: 95-96°/5mm Beilstein Reference: 2650701 MDL No.: MFCD00014706
Formel: C₇H₃BrF₃NO₂
MW: 270,01 g/mol
Kokpunkt: 95…96 °C (5 mmHg)
Smältpunkt: 32…35 °C
Densitet: 1,799
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00014706
CAS nummer: 344-38-7