ω-Bromo-2-methoxyacetophenone ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: 2-Methoxyphenacyl bromide, α-Bromo-2-methoxyacetophenone, 2-Bromo-2'-methoxyacetophenone, 2-Bromo-1-(2-methoxyphenyl)ethanone, 2-Bromo-1-(2-methoxyphenyl)ethan-1-one, Bromomethyl o-methoxyphenyl ketone
L07696.03 L07696.06
L07696.03EA 795 SEK
L07696.03 L07696.06
ω-Bromo-2-methoxyacetophenone ≥98%
ω-Bromo-2-methoxyacetophenone
Purity: 98% CAS No.: 31949-21-0 Molecular Formula: C9H9BrO2 Formula Weight: 229.08 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 42-46° Boiling Point: 130°/1mm Beilstein Reference: 1938788 UN No.: 3261 MDL No.: MFCD00000196
Formel: C₉H₉BrO₂
MW: 229,07 g/mol
Kokpunkt: 130 °C (1 mmHg)
Smältpunkt: 42…46 °C
Flampunkt: >110 °C (230 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00000196
CAS nummer: 31949-21-0
EINECS: 250-870-2
UN: 3261
ADR: 8,III