4'-Bromoacetophenone ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: p-Bromoacetophenone, 1-(4-Bromophenyl)ethanone
A12222.14 A12222.22 A12222.36
A12222.14EA 525 SEK
A12222.14 A12222.22 A12222.36
4'-Bromoacetophenone ≥98%
4'-Bromoacetophenone
Purity: 98% CAS No.: 99-90-1 Molecular Formula: C8H7BrO Formula Weight: 199.05 Flash Point: 110°(230°F) Melting Point: 49-53° Boiling Point: 255-256° Beilstein Reference: 386015 Merck Reference: 14,1403 MDL No.: MFCD00000105
Formel: C₈H₇BrO
MW: 199,05 g/mol
Kokpunkt: 255…256 °C
Smältpunkt: 49…53 °C
Densitet: 1,736 g/cm³ (20 °C)
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000105
CAS nummer: 99-90-1
EINECS: 202-799-3
UN: 2811
ADR: 6.1,II
Merck Index: 14,1403