3'-Bromoacetophenone 97%
Leverantör: Thermo Scientific
Varning
Synonyms: m-Bromoacetophenone
106580250 106580050 106581000
ACRO106580250EA 740 SEK
ACRO106580250 ACRO106580050 ACRO106581000
3'-Bromoacetophenone 97%
3'-Bromoacetophenone
Formel: C₈H₇BrO
MW: 199,05 g/mol
Kokpunkt: 79…81 °C (2 mmHg)
Densitet: 1,49 g/cm³
Flampunkt: >112 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000083
CAS nummer: 2142-63-4
EINECS: 218-396-0
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5749 to 1.5769 (20°C, 589 nm)
GC ≥96.0 %
Residue after ignition ≤0.3 %
Specific optical rotation -45° to -51° (20°C, 589 nm) (c=10, 75% acetic acid)