2'-Bromoacetophenone ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: o-Bromoacetophenone
A11982.09 A11982.18 A11982.30
A11982.09EA 1225 SEK
A11982.09 A11982.18 A11982.30
2'-Bromoacetophenone ≥99%
2'-Bromoacetophenone
Purity: 99% CAS No.: 2142-69-0 Molecular Formula: C8H7BrO Formula Weight: 199.05 Flash Point: 110°(230°F) Refractive Index: 1.5680 Boiling Point: 116-117°/12mm Beilstein Reference: 1931534 MDL No.: MFCD00000067
Formel: C₈H₇BrO
MW: 199,05 g/mol
Kokpunkt: 116…117 °C (12 mmHg)
Smältpunkt: 0 °C
Densitet: 1,476
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000067
CAS nummer: 2142-69-0
EINECS: 218-398-1