α-Bromo-3,4-dichlorotoluene

Leverantör: Thermo Scientific
Fara

Synonyms: 3,4-Dichlorobenzyl bromide

SB01686DA SB01686EA
MAYBSB01686DAEA 0 SEK
MAYBSB01686DA MAYBSB01686EA
α-Bromo-3,4-dichlorotoluene
α-Bromo-3,4-dichlorotoluene
Formel: C₇H₅BrCl₂
MW: 239.93 g/mol
Kokpunkt: 84…86 °C (0.3 mmHg)
MDL Number: MFCD00041363
CAS nummer: 18880-04-1
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR