2-Formylfenylboronsyra 98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 2-Formylbenzeneboronic acid, (2-Formylphenyl)metaboric acid, 2-Boronobenzaldehyde, (2-Formylphenyl)boronic acid, Dihydroxy(2-formylphenyl)borane
359360010 359360050
ACRO359360010EA 835 SEK
ACRO359360010 ACRO359360050
2-Formylfenylboronsyra 98%
2-Formylfenylboronsyra
Formel: C₇H₇BO₃
MW: 149.94 g/mol
Smältpunkt: 120…122 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00151822
CAS nummer: 40138-16-7
Order Now
Specifikation testresultat
Appearance White to pink to orange Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %