Phenylacetylene 98%, ren

Leverantör: Thermo Scientific
Fara

152460250 152461000 152465000
ACRO152460250EA 755 SEK
ACRO152460250 ACRO152461000 ACRO152465000
Phenylacetylene 98%, ren
Phenylacetylene
Formel: C₈H₆
MW: 102,14 g/mol
Kokpunkt: 142…144 °C (760 mmHg)
Smältpunkt: –44,8 °C
Densitet: 0,93 g/cm³
Flampunkt: 31 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00008570
CAS nummer: 536-74-3
EINECS: 208-645-1
UN: 3295
ADR: 3,III
Merck Index: 13,03892

Order Now

Specifikation testresultat

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5470 to 1.5500 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR