1-Fluoro-4-ethynylbenzene 98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1-Ethynyl-4-fluorobenzene, (4-Fluorophenyl)acetylene, (4-Fluorophenyl)ethyne
371370010 371370050
ACRO371370010EA 985 SEK
ACRO371370010 ACRO371370050
1-Fluoro-4-ethynylbenzene 98%
1-Fluoro-4-ethynylbenzene
Formel: C₈H₅F
Kokpunkt: 55…56 °C (40 mmHg)
Smältpunkt: 27…28 °C
Densitet: 1,048 g/cm³
Flampunkt: 29 °C
Storage Temperature: Omgivande
MDL Number: MFCD00168823
CAS nummer: 766-98-3
UN: 1325
ADR: 4.1,III
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5155 to 1.5185 (20°C, 589 nm)
GC ≥97.5 %