Resveratrol
Leverantör: Cayman Chemical
Synonyms: trans-3,4,5-Trihydroxystilbene, (E)-5-(4-Hydroxystyryl)benzene-1,3-diol
70675-100 70675-50 70675-500 70675-250
CAYM70675-100EA 1325 SEK
CAYM70675-100 CAYM70675-50 CAYM70675-500 CAYM70675-250
Resveratrol
Resveratrol
Formel: C₁₄H₁₂O₃
MW: 228,25 g/mol
Smältpunkt: 260 °C
Densitet: 1,36 g/cm³ (20 °C)
Storage Temperature: Frys
MDL Number: MFCD00133799
CAS nummer: 501-36-0
Merck Index: 12,08326