2-Allylphenol ≥98%

Leverantör: Thermo Fisher Scientific
Fara

A10755.22 A10755.36
A10755.22EA 900 SEK
A10755.22 A10755.36
2-Allylphenol ≥98%
2-Allylphenol

Purity: 98+% CAS No.: 1745-81-9 Molecular Formula: C9H10O Formula Weight: 134.18 Flash Point: 88°(190°F) Refractive Index: 1.5450 Odour: Phenol-like Melting Point: -6° Boiling Point: 220-222° Beilstein Reference: 742121 UN No.: 2922 MDL No.: MFCD00002250

Formel: C₉H₁₀O
Kokpunkt: 220…222 °C
Smältpunkt: –6 °C
Densitet: 1,024
Flampunkt: 88 °C (190 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00002250
CAS nummer: 1745-81-9
EINECS: 217-119-0
UN: 2922
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR