4-Allyl-2,6-dimethoxyphenol ≥98%
Leverantör: Thermo Fisher Scientific
Varning
B21981.03 B21981.06 B21981.14
B21981.03EA 1100 SEK
B21981.03 B21981.06 B21981.14
4-Allyl-2,6-dimethoxyphenol ≥98%
4-Allyl-2,6-dimethoxyphenol
Purity: 98% CAS No.: 6627-88-9 Molecular Formula: C11H14O3 Formula Weight: 194.23 Flash Point: 110°(230°F) Refractive Index: 1.5480 Boiling Point: 168-169°/11mm MDL No.: MFCD00008655
Formel: C₁₁H₁₄O₃
MW: 194.23 g/mol
Kokpunkt: 168…169 °C (11 mmHg)
Densitet: 1.092
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008655
CAS nummer: 6627-88-9
EINECS: 229-600-2