Eugenol ≥99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 4-Allyl-2-methoxyphenol, 5-Allyl-2-hydroxyanisole
A14332.22 A14332.36
A14332.22EA 870 SEK
A14332.22 A14332.36
Eugenol ≥99%
Eugenol
Purity: 99% CAS No.: 97-53-0 Molecular Formula: C10H12O2 Formula Weight: 164.21 Flash Point: 127°(260°F) Refractive Index: 1.5410 Odour: Pungent Melting Point: -10° Boiling Point: 252-253° Beilstein Reference: 1366759 Merck Reference: 14,3898 MDL No.: MFCD00008654
Formel: C₁₀H₁₂O₂
MW: 164.2 g/mol
Kokpunkt: 252…253 °C
Smältpunkt: –10 °C
Densitet: 1.068
Flampunkt: 127 °C (260 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008654
CAS nummer: 97-53-0
EINECS: 202-589-1
Merck Index: 13,03931