4-Allylanisole 98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1-Allyl-4-methoxybenzene, Estragole
102880250 102881000 102880050
ACRO102880250EA 980 SEK
ACRO102880250 ACRO102881000 ACRO102880050
4-Allylanisole 98%
4-Allylanisole
Formel: C₁₀H₁₂O
MW: 148.2 g/mol
Kokpunkt: 216
Smältpunkt: 22.5
Densitet: 0.96 g/cm³
Flampunkt: 81 °C
MDL Number: MFCD00008653
CAS nummer: 140-67-0
EINECS: 205-427-8
Merck Index: 13,03740
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5190 to 1.5240 (20°C, 589 nm)
GC ≥97.5 %
Specific optical rotation +20.5° to +21.5° (25°C, 589 nm) (c=10, H2O)
Iron (Fe) ≤10 ppm
Heavy metals (ICP-OES) ≤20 ppm