2,4-Dinitrobenzenesulphonyl chloride ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: 2,4-Dinitrobenzene-1-sulphonyl chloride
B23035.06 B23035.14 B23035.22
B23035.06EA 1300 SEK
B23035.06 B23035.14 B23035.22
2,4-Dinitrobenzenesulphonyl chloride ≥98%
2,4-Dinitrobenzenesulphonyl chloride
Purity: 98% CAS No.: 1656-44-6 Molecular Formula: C6H3ClN2O6S Formula Weight: 266.62 Melting Point: 100-104° UN No.: 3096 MDL No.: MFCD00007431
Formel: C₆H₃ClN₂O₆S
MW: 266,62 g/mol
Smältpunkt: 100…104 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007431
CAS nummer: 1656-44-6
EINECS: 216-749-3
UN: 3261
ADR: 8,II