o-Fenylendiamin 98%
Leverantör: Thermo Scientific
Fara
Synonyms: 1,2-Fenylendiamin
130550010 130552500 130550050
ACRO130550010EA 3625 SEK
ACRO130550010 ACRO130552500 ACRO130550050
o-Fenylendiamin 98%
o-Fenylendiamin
Formel: C₆H₈N₂
MW: 108,14 g/mol
Kokpunkt: 256…258 °C (760 mmHg)
Smältpunkt: 100…103 °C
Densitet: 1,21 g/cm³ (20 °C)
Flampunkt: 136 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007721
CAS nummer: 95-54-5
EINECS: 202-430-6
UN: 1673
ADR: 6.1,III
Merck Index: 12,07438
Order Now
Specifikation testresultat
Appearance Tan to brown Crystalline powder or crystals or flakes (May darken on storage)
Infrared spectrum Conforms
Melting point 100 to 104 °C
GC ≥97.5 %
Residue after evaporation ≤0.001 %