Sulfanilinsyra 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 4-Aminobenzensulfonsyra
150710010 150711000 150710025 150710100
ACRO150710010EA 1675 SEK
ACRO150710010 ACRO150711000 ACRO150710025 ACRO150710100
Sulfanilinsyra 99%
Sulfanilinsyra
Formel: 4-(H₂N)C₆H₄SO₃H
MW: 173,19 g/mol
Kokpunkt: 363 °C (1013 hPa)
Smältpunkt: 365 °C
Densitet: 1,485 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007886
CAS nummer: 121-57-3
EINECS: 204-482-5
Merck Index: 13,09012
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e032bed891f24ee48419dc3c7237b0293fd3e629&vid=4105810d5bf1feff3214fe3bfcdd8d1ae1443204&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Off-white to beige to blue-grey Powder
Infrared spectrum Conforms
Titration with NaOH ≥98.5 %