3-Aminothiophenol 97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 3-Aminophenyl mercaptan, 3-Mercaptoaniline, 3-Aminobenzenethiol
B25473.06 B25473.14
B25473.06EA 1850 SEK
B25473.06 B25473.14
3-Aminothiophenol 97%
3-Aminothiophenol
Purity: 97% CAS No.: 22948-02-3 Molecular Formula: C6H7NS Formula Weight: 125.19 Flash Point: 100°(212°F) Refractive Index: 1.6590 Boiling Point: 123-125°/8mm UN No.: 3267 MDL No.: MFCD00007791
Formel: C₆H₇NS
Kokpunkt: 123…125 °C (8 mmHg)
Densitet: 1,179
Flampunkt: >100 °C (212 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00007791
CAS nummer: 22948-02-3
EINECS: 245-344-4
UN: 3267
ADR: 8,III