3-Aminofenol 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 3-Hydroxyaniline, m-Aminophenol
146102500 146100010 146100050
ACRO146102500EA 950 SEK
ACRO146102500 ACRO146100010 ACRO146100050
3-Aminofenol 99%
3-Aminofenol
Formel: C₆H₇NO
MW: 109,13 g/mol
Kokpunkt: 164 °C (15hPa)
Smältpunkt: 121…124 °C
Densitet: 1,275 g/cm³ (20 °C)
Flampunkt: 155 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007786
CAS nummer: 591-27-5
EINECS: 209-711-2
UN: 2512
ADR: 6.1,III
Merck Index: 13,00459
Order Now
Specifikation testresultat
Appearance White to grey to light brown Crystalline powder or crystals or flakes
Infrared spectrum Conforms
Melting point 120 to 124 °C
GC ≥98.5 %