p-Anisidin 99
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 4-Metoxianilin, 4-Anisidin
A10946.0B A10946.22 A10946.36
A10946.0BEA 3375 SEK
A10946.0B A10946.22 A10946.36
p-Anisidin 99
p-Anisidin
Purity: 99% CAS No.: 104-94-9 Molecular Formula: C7H9NO Formula Weight: 123.16 Flash Point: 122°(251°F) Melting Point: 56-60° Boiling Point: 240-243° Beilstein Reference: 471556 Merck Reference: 14,667 UN No.: 2811 MDL No.: MFCD00007864
Formel: C₇H₉NO
MW: 123.15 g/mol
Kokpunkt: 240…243 °C
Smältpunkt: 56…60 °C
Densitet: 1.089
Flampunkt: 122 °C (251 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00007864
CAS nummer: 104-94-9
EINECS: 203-254-2
UN: 2811
ADR: 6.1 III
Merck Index: 13,00670