m-Anisidin ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 3-Anisidin, 3-Metoxianilin
A10815.0B A10815.18 A10815.30
A10815.0BEA 6975 SEK
A10815.0B A10815.18 A10815.30
m-Anisidin ≥98%
m-Anisidin
Purity: 98% CAS No.: 536-90-3 Molecular Formula: C7H9NO Formula Weight: 123.16 Flash Point: 126°(258°F) Refractive Index: 1.5810 Odour: Characteristic Melting Point: -1° Boiling Point: 250-251° Beilstein Reference: 386119 Merck Reference: 14,667 UN No.: 2431 MDL No.: MFCD00007783
Formel: C₇H₉NO
MW: 123.15 g/mol
Kokpunkt: 250…251 °C
Smältpunkt: –1 °C
Densitet: 1.101
Flampunkt: 126 °C (258 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00007783
CAS nummer: 536-90-3
EINECS: 208-651-4
UN: 2431
ADR: 6.1 III
Merck Index: 13,00670