4-Methoxyphenyl isothiocyanate

Leverantör: Thermo Scientific

Synonyms: 4-Methoxyphenylisothiocyanate

TL00326EA
MAYBTL00326EAEA 0 SEK
MAYBTL00326EA
4-Methoxyphenyl isothiocyanate
4-Methoxyphenyl isothiocyanate
Formel: C₈H₇NOS
MW: 165,22 g/mol
Kokpunkt: 279…281 °C (1013 hPa)
Smältpunkt: 17…19 °C
Densitet: 1,18 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011676
CAS nummer: 2284-20-0
EINECS: 218-921-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR