4-Methoxyphenyl isothiocyanate ≥98%
Leverantör: Thermo Fisher Scientific
Fara
A15331.04 A15331.09 A15331.18
A15331.04EA 800 SEK
A15331.04 A15331.09 A15331.18
4-Methoxyphenyl isothiocyanate ≥98%
4-Methoxyphenyl isothiocyanate
Purity: 98% CAS No.: 2284-20-0 Molecular Formula: C8H7NOS Formula Weight: 165.22 Flash Point: 110°(230°F) Refractive Index: 1.6489 Odour: Pungent Melting Point: 18-20° Boiling Point: 279-281° Beilstein Reference: 606967 UN No.: 2922 MDL No.: MFCD00011676
Formel: C₈H₇NOS
Kokpunkt: 279…281 °C
Smältpunkt: 18…20 °C
Densitet: 1,196
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00011676
CAS nummer: 2284-20-0
EINECS: 218-921-3
ADR: 8,III