2-Methoxyphenyl isothiocyanate

TL00631DA
MAYBTL00631DAEA 0 SEK
MAYBTL00631DA
2-Methoxyphenyl isothiocyanate
2-Methoxyphenyl isothiocyanate
Formel: C₈H₇NOS
MW: 165,22 g/mol
MDL Number: MFCD00011675
CAS nummer: 3288-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR