2-Methoxyphenyl isothiocyanate ≥98%
Leverantör: Thermo Fisher Scientific
Fara
L11799.06 L11799.14
L11799.06EA 1675 SEK
L11799.06 L11799.14
2-Methoxyphenyl isothiocyanate ≥98%
2-Methoxyphenyl isothiocyanate
Purity: 98% CAS No.: 3288-04-8 Molecular Formula: C8H7NOS Formula Weight: 165.22 Flash Point: 110°(230°F) Refractive Index: 1.6443 Odour: Pungent Boiling Point: 267° Beilstein Reference: 638293 UN No.: 2922 MDL No.: MFCD00011675
Formel: C₈H₇NOS
MW: 165.22 g/mol
Kokpunkt: 267 °C
Densitet: 1.196
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00011675
CAS nummer: 3288-04-8
ADR: 8 III