2-Nitrophenyl isocyanate 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Isocyanato-2-nitrobenzene, 2-Nitrophenylisocyanate
296690050 296690250 AC29669EA
ACRO296690050EA 1225 SEK
ACRO296690050 ACRO296690250 MAYBAC29669EA
2-Nitrophenyl isocyanate 99%
2-Nitrophenyl isocyanate
Formel: C₇H₄N₂O₃
MW: 164,12 g/mol
Kokpunkt: 135…137 °C (17 mmHg)
Smältpunkt: 38…41 °C
Flampunkt: >110 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00007092
CAS nummer: 3320-86-3
EINECS: 222-024-2
UN: 3335
ADR: 9,III
Order Now
Specifikation testresultat
Appearance Yellow Crystalline solid
Infrared spectrum Conforms
Melting point 38 to 41 °C
GC ≥98.5 %