N,N-Dimetyl-1,4-benzendiamin 97%
Leverantör: Thermo Scientific
Fara
Synonyms: N,N-Dimetyl-p-benzendiamin
408460050 408460250 408461000 408465000
ACRO408460050EA 665 SEK
ACRO408460050 ACRO408460250 ACRO408461000 ACRO408465000
N,N-Dimetyl-1,4-benzendiamin 97%
N,N-Dimetyl-1,4-benzendiamin
Formel: C₈H₁₂N₂
MW: 136,2 g/mol
Kokpunkt: 262 °C (1013 hPa)
Smältpunkt: 35 °C
Densitet: 1,09 g/cm³
Flampunkt: 130 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007860
CAS nummer: 99-98-9
EINECS: 202-807-5
UN: 2811
ADR: 6.1,II
Merck Index: 13,33280
Order Now
Specifikation testresultat
Appearance Dark brown to black Fused mass or liquid after melting
Infrared spectrum Conforms
GC ≥96.0 %