3-Amino-2,4,6-triiodobenzoic acid, (max. 1 mol H₂O) 99%
Leverantör: Thermo Fisher Scientific
Varning
B23405.06 B23405.14
B23405.06EA 1250 SEK
B23405.06 B23405.14
3-Amino-2,4,6-triiodobenzoic acid, (max. 1 mol H₂O) 99%
3-Amino-2,4,6-triiodobenzoic acid
Purity: 99% CAS No.: 3119-15-1 Molecular Formula: C7H4I3NO2 Formula Weight: 514.83 Melting Point: ca 196° dec. MDL No.: MFCD00007681
Formel: C₇H₄I₃NO₂
MW: 514.83 g/mol
Kokpunkt: 459
Smältpunkt: ∼ 196 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00007681
CAS nummer: 3119-15-1
EINECS: 221-493-0