Methyl-4-iodobenzoate ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 4-Iodobenzoic acid methyl ester, Methyl 4-iodobenzoate
A19342.09 A19342.18 A19342.30
A19342.09EA 985 SEK
A19342.09 A19342.18 A19342.30
Methyl-4-iodobenzoate ≥98%
Methyl-4-iodobenzoate
Purity: 98% CAS No.: 619-44-3 Molecular Formula: C8H7IO2 Formula Weight: 262.05 Melting Point: 114-116° MDL No.: MFCD00016353
Formel: C₈H₇IO₂
MW: 262.05 g/mol
Kokpunkt: 276
Smältpunkt: 114…116 °C
Densitet: 2.02 g/cm³ (10 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00016353
CAS nummer: 619-44-3
EINECS: 210-597-1