Ethyl 2-iodobenzoate ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Ethyl 2-iodobenzoate, 2-Iodobenzoic acid ethyl ester
L07650.14 L07650.06
L07650.14EA 3525 SEK
L07650.14 L07650.06
Ethyl 2-iodobenzoate ≥98%
Ethyl-2-iodobenzoate
Purity: 98% CAS No.: 1829-28-3 Molecular Formula: C9H9IO2 Formula Weight: 276.08 Refractive Index: 1.5840 Boiling Point: 163-165°/23mm MDL No.: MFCD00051796
Formel: C₉H₉IO₂
MW: 276.07 g/mol
Kokpunkt: 163…165 °C (23 mmHg)
Storage Temperature: Omgivande
MDL Number: MFCD00051796
CAS nummer: 1829-28-3