3,4-Difluorobenzenesulphonyl chloride ≥97%
Leverantör: Thermo Scientific
Fara
Synonyms: 3,4-Difluorobenzene-1-sulphonyl chloride
L17816.03 L17816.06
L17816.03EA 1025 SEK
L17816.03 L17816.06
3,4-Difluorobenzenesulphonyl chloride ≥97%
3,4-Difluorobenzenesulphonyl chloride
Purity: 97% CAS No.: 145758-05-0 Molecular Formula: C6H3ClF2O2S Formula Weight: 212.60 Refractive Index: 1.5140 Boiling Point: 105°/15mm UN No.: 3265 MDL No.: MFCD00153120
Formel: C₆H₃ClF₂O₂S
MW: 212,6 g/mol
Kokpunkt: 105 °C (15 mmHg)
Densitet: 1,586
Storage Temperature: Omgivande
MDL Number: MFCD00153120
CAS nummer: 145758-05-0
UN: 3265
ADR: 8,II