2,4-Difluorobenzenesulphonyl chloride ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: 2,4-Difluorobenzene-1-sulphonyl chloride
A14639.06 A14639.14 A14639.22
A14639.06EA 1550 SEK
A14639.06 A14639.14 A14639.22
2,4-Difluorobenzenesulphonyl chloride ≥98%
2,4-Difluorobenzenesulphonyl chloride
Purity: 98% CAS No.: 13918-92-8 Molecular Formula: C6H3ClF2O2S Formula Weight: 212.60 Flash Point: 110°(230°F) Refractive Index: 1.5187 Boiling Point: 236-237° Beilstein Reference: 2970373 UN No.: 3265 MDL No.: MFCD00042481
Formel: C₆H₃ClF₂O₂S
MW: 212,6 g/mol
Kokpunkt: 236…237 °C
Densitet: 1,581
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00042481
CAS nummer: 13918-92-8
UN: 3265
ADR: 8,II