4-(Trifluoromethoxy)benzenesulphonyl chloride ≥98%
Leverantör: Thermo Fisher Scientific
Fara
A15467.03 A15467.06 A15467.14
A15467.03EA 610 SEK
A15467.03 A15467.06 A15467.14
4-(Trifluoromethoxy)benzenesulphonyl chloride ≥98%
4-(Trifluoromethoxy)benzenesulphonyl chloride
Purity: 98% CAS No.: 94108-56-2 Molecular Formula: C7H4ClF3O3S Formula Weight: 260.62 Flash Point: 110°(230°F) Refractive Index: 1.4800 Boiling Point: 120-122°/15mm Beilstein Reference: 7641524 UN No.: 3265 MDL No.: MFCD00042408
Formel: C₇H₄ClF₃O₃S
Kokpunkt: 120…122 °C (15 mmHg)
Densitet: 1,536
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00042408
CAS nummer: 94108-56-2
EINECS: 302-397-9
UN: 3265
ADR: 8,II