2,4-Difluoroanisole ≥99.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2,4-Difluoro-1-methoxybenzene

D1849-10G D1849-25G
TCIAD1849-10GEA 615 SEK
TCIAD1849-10G TCIAD1849-25G
2,4-Difluoroanisole ≥99.0%
2,4-Difluoroanisole
Formel: C₇H₆F₂O
MW: 144,12 g/mol
Kokpunkt: 53°C
Densitet: 1,2513 g/cm³ (20.7 °C)
Flampunkt: 51°C
MDL Number: MFCD00010731
CAS nummer: 452-10-8
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73112d9e07e5fd15fb49a73b1cdb9e4b64bb53ce&vid=65d26373585cfefaa944daa1a7e1eac6605c2539&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR