2-(Trifluoromethoxy)aniline
Leverantör: Thermo Scientific
SB01054EE
MAYBSB01054EEEA
0
SEK
MAYBSB01054EE
2-(Trifluoromethoxy)aniline
2-(Trifluoromethoxy)aniline
Formel:
C₇H₆F₃NO MW: 177,13 g/mol Kokpunkt: 64,5 °C (21 mmHg) Densitet: 1,301 g/cm³ (20 °C) |
MDL Number:
MFCD00035959 CAS nummer: 1535-75-7 EINECS: 216-257-9 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...